Learn More
2-Cyano-6-hydroxybenzothiazole, 97%, Thermo Scientific Chemicals
122.61 € - 492.00 €
Chemical Identifiers
CAS | 939-69-5 |
---|---|
Molecular Formula | C8H4N2OS |
Molecular Weight (g/mol) | 176.19 |
MDL Number | MFCD00296905 |
InChI Key | SQAVNBZDECKYOT-UHFFFAOYSA-N |
Synonym | 2-cyano-6-hydroxybenzothiazole, 6-hydroxybenzo d thiazole-2-carbonitrile, 6-hydroxybenzothiazole-2-carbonitrile, 2-benzothiazolecarbonitrile, 6-hydroxy, 6-hydroxy-2-benzothiazolecarbonitrile, pubchem19680, 6-hydroxy-2-cyanobenzothiazole, 6-hydroxy-2-cyano-benzothiazole, 6-hydroxy-benzothiazole-2-carbonitrile, 6-hydroxy-benzo d thiazole-2-carbonitrile |
PubChem CID | 9881912 |
IUPAC Name | 6-hydroxy-1,3-benzothiazole-2-carbonitrile |
SMILES | OC1=CC=C2N=C(SC2=C1)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15256018
|
Thermo Scientific Acros
450940010 |
1 g |
122.61 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15266018
|
Thermo Scientific Acros
450940050 |
5 g |
492.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
939-69-5 | |
176.19 | |
SQAVNBZDECKYOT-UHFFFAOYSA-N | |
9881912 | |
OC1=CC=C2N=C(SC2=C1)C#N |
C8H4N2OS | |
MFCD00296905 | |
2-cyano-6-hydroxybenzothiazole, 6-hydroxybenzo d thiazole-2-carbonitrile, 6-hydroxybenzothiazole-2-carbonitrile, 2-benzothiazolecarbonitrile, 6-hydroxy, 6-hydroxy-2-benzothiazolecarbonitrile, pubchem19680, 6-hydroxy-2-cyanobenzothiazole, 6-hydroxy-2-cyano-benzothiazole, 6-hydroxy-benzothiazole-2-carbonitrile, 6-hydroxy-benzo d thiazole-2-carbonitrile | |
6-hydroxy-1,3-benzothiazole-2-carbonitrile |
Specifications
939-69-5 | |
Authentic | |
Glass Bottle | |
MFCD00296905 | |
SQAVNBZDECKYOT-UHFFFAOYSA-N | |
6-hydroxy-1,3-benzothiazole-2-carbonitrile | |
9881912 | |
97% |
212.0°C to 215.0°C | |
96% min. (HPLC) | |
C8H4N2OS | |
2-cyano-6-hydroxybenzothiazole, 6-hydroxybenzo d thiazole-2-carbonitrile, 6-hydroxybenzothiazole-2-carbonitrile, 2-benzothiazolecarbonitrile, 6-hydroxy, 6-hydroxy-2-benzothiazolecarbonitrile, pubchem19680, 6-hydroxy-2-cyanobenzothiazole, 6-hydroxy-2-cyano-benzothiazole, 6-hydroxy-benzothiazole-2-carbonitrile, 6-hydroxy-benzo d thiazole-2-carbonitrile | |
OC1=CC=C2N=C(SC2=C1)C#N | |
176.19 | |
176.2 | |
2-Cyano-6-hydroxybenzothiazole |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
- 2-Cyano-6-hydroxybenzothiazole, 97%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only