missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyanothioacetamide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.
Solubility
Does not mix well with water.
Notes
Light Sensitive. Store in the dark.
2-Cyanothioacetamide was used in the synthesis of 3,4-trans-4-aryl-3-(1-pyridinio)-1,2,3,4-tetrahydropyridine-6-thiolates and bis[6-(2-aryl)-2-thioxo-1,2-dihydropyridine-3-carbonitrile]. 2-Cyanothioacetamide is used as a building block for 2-pyridothiones. It can be used in agrochemical, pharmaceutical and dyestuff field etc,.
Solubility
Does not mix well with water.
Notes
Light Sensitive. Store in the dark.
Specifications
Specifications
Melting Point | 114°C to 120°C |
Quantity | 5 g |
UN Number | UN3439 |
Beilstein | 1699934 |
Sensitivity | Light sensitive |
Solubility Information | Does not mix well with water. |
Formula Weight | 100.14 |
Percent Purity | 98% |
Chemical Name or Material | 2-Cyanothioacetamide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'