Learn More
2-Cyclohexen-1-one, 97%, Thermo Scientific Chemicals
45.53 € - 475.37 €
Chemical Identifiers
CAS | 930-68-7 |
---|---|
Molecular Formula | C6H8O |
Molecular Weight (g/mol) | 96.13 |
MDL Number | MFCD00001577 |
InChI Key | FWFSEYBSWVRWGL-UHFFFAOYSA-N |
Synonym | 2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one |
PubChem CID | 13594 |
ChEBI | CHEBI:15977 |
IUPAC Name | cyclohex-2-en-1-one |
SMILES | O=C1CCCC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10112380
|
Thermo Scientific Acros
111240100 |
10 g | Glass bottle |
45.53 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645912
|
Thermo Scientific Acros
111240250 |
25 g | Glass bottle |
62.11 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10395690
|
Thermo Scientific
111241000 |
100 g | Glass bottle |
216.08 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984831
|
Thermo Scientific Acros
111245000 |
500 g | Glass bottle |
475.37 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
930-68-7 | |
96.13 | |
FWFSEYBSWVRWGL-UHFFFAOYSA-N | |
13594 | |
cyclohex-2-en-1-one |
C6H8O | |
MFCD00001577 | |
2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one | |
CHEBI:15977 | |
O=C1CCCC=C1 |
Specifications
930-68-7 | |
100.0 | |
Yellow | |
168.0°C | |
Authentic | |
C6H8O | |
1.4875 to 1.4895 | |
07,II,55 | |
0.993 | |
Solubility in water: soluble. Other solubilities: soluble in organic solvents | |
O=C1CCCC=C1 | |
96.13 | |
CHEBI:15977 | |
97% | |
2-Cyclohexen-1-one |
96.0 | |
-53.0°C | |
0.9930g/mL | |
59°C | |
97% | |
C6H8(=O) | |
MFCD00001577 | |
01,18 | |
2-cyclohexen-1-one, cyclohex-2-enone, 2-cyclohexenone, cyclohexenone, 3-oxocyclohexene, 1-cyclohexen-3-one, cyclohexen-3-one, cyclohexen-1-one, 2-cyclohexenone-1, 2-cyclohexene-1-one | |
FWFSEYBSWVRWGL-UHFFFAOYSA-N | |
cyclohex-2-en-1-one | |
13594 | |
96.13 | |
Liquid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Fatal in contact with skin.
Fatal if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiou
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-223-5
- 2-Cyclohexen-1-one
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
Hazard Statement
- H310-Fatal in contact with skin.
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.