missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclohexylethanol, 99%, Thermo Scientific Chemicals

Product Code. 11475676
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a flavoring agent.

Solubility
Soluble in alcohol and dipropylene glycol. Soluble in water (1750 mg/L at 25°C).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4442-79-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001525
InChI Key QJQZRLXDLORINA-UHFFFAOYSA-N
Synonym cyclohexaneethanol, cyclohexylethanol, cyclohexylethyl alcohol, 2-cyclohexylethan-1-ol, ethanol, 2-cyclohexyl, hexahydrophenylethyl alcohol, unii-653aok9puo, 2-hydroxyethyl cyclohexane, .beta.-cyclohexylethyl alcohol, 2-cyclohexyl-ethanol
PubChem CID 20508
IUPAC Name 2-cyclohexylethanol
SMILES C1CCC(CC1)CCO

Specifications

Melting Point -20°C
Density 0.92
Boiling Point 206°C to 207°C
Flash Point 86°C (186°F)
Refractive Index 1.466
Quantity 250 g
Beilstein 1848152
Solubility Information Soluble in alcohol and dipropylene glycol. Soluble in water (1750mg/L at 25°C).
Formula Weight 128.22
Percent Purity 99%
Chemical Name or Material 2-Cyclohexylethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'