missing translation for 'onlineSavingsMsg'
Learn More

2-Cyclopentenone, 98%

Product Code. p-3736080 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10224090

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 930-30-3 | C5H6O | 82.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-30-3
Molecular Formula C5H6O
Molecular Weight (g/mol) 82.10
MDL Number MFCD00001401
InChI Key BZKFMUIJRXWWQK-UHFFFAOYSA-N
Synonym 2-cyclopenten-1-one, 2-cyclopentenone, cyclopentenone, cyclopent-2-enone, cyclopenten-3-one, 2-cyclopentene-1-one, cyclopent-2-ene-1-one, unii-q0u2igf9ck, 3-cyclopenten-2-one, q0u2igf9ck
PubChem CID 13588
IUPAC Name cyclopent-2-en-1-one
SMILES O=C1CCC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.9800g/mL
Boiling Point 150°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C5H6(=O)
Refractive Index 1.4800 to 1.482
Quantity 5 g
Beilstein 07, I, 45
Specific Gravity 0.98
Solubility Information Solubility in water: almost insoluble. Other solubilities: very soluble in ethanol,ether
Formula Weight 82.1
Percent Purity 98%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Cyclopentenone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Cyclopentenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.