missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Cyclopropylethanol, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
2-Cyclopropylethanol is used to prepare (2-bromo-ethyl)-cyclopropane by reacting with phosphorus tribromide.
Solubility
Miscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
Specifications
Boiling Point | 137°C to 138°C |
Flash Point | 47°C (116°F) |
Refractive Index | 1.4355 |
Quantity | 250 mg |
UN Number | UN1987 |
Beilstein | 2036028 |
Solubility Information | Miscible with water. |
IUPAC Name | 2-cyclopropylethanol |
Formula Weight | 86.13 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'