Learn More
Thermo Scientific Chemicals 2'-Deoxycytidine-5'-monophosphate, 99%
Description
2'-Deoxycytidine-5'-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2′-Deoxycytidine-5′-monophosphate is used as a substrate of uridine monophosphate (UMP)/cytidine monophosphate (CMP) kinase (EC 2.7.4.4) to form dCDP which upon phosphorylation to dCTP supports DNA and RNA biosynthesis.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Color | White |
Quantity | 1 g |
Beilstein | 41062 |
Solubility Information | Soluble in water. |
Formula Weight | 307.2 |
Percent Purity | 99% |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2'-Deoxycytidine-5'-monophosphate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.