Learn More
2-(Diethylamino)ethyl chloride hydrochloride, 98+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 869-24-9 |
---|---|
Molecular Formula | C6H15Cl2N |
Molecular Weight (g/mol) | 172.09 |
MDL Number | MFCD00012519 |
InChI Key | RAGSWDIQBBZLLL-UHFFFAOYSA-N |
Synonym | 2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride |
PubChem CID | 13363 |
SMILES | [H+].[Cl-].CCN(CC)CCCl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
869-24-9 | |
172.09 | |
RAGSWDIQBBZLLL-UHFFFAOYSA-N | |
13363 |
C6H15Cl2N | |
MFCD00012519 | |
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride | |
[H+].[Cl-].CCN(CC)CCCl |
Specifications
869-24-9 | |
5 | |
(CH3CH2)2NCH2CH2Cl·HCl | |
UN2928 | |
2-chloro-n,n-diethylethanamine hydrochloride, 2-diethylaminoethylchloride hydrochloride, 2-chlorotriethylamine hydrochloride, 2-chloro-n,n-diethylethylamine hydrochloride, 2-diethylamino ethyl chloride hydrochloride, diethylaminoethyl chloride hydrochloride, n,n-diethyl-2-chloroethylamine hydrochloride, 2-chloroethyl diethylamine hydrochloride, unii-ycy6eqt3wa, ethanamine, 2-chloro-n,n-diethyl-, hydrochloride | |
[H+].[Cl-].CCN(CC)CCCl | |
172.09 | |
172.1 | |
2-(Diethylamino)ethyl chloride hydrochloride |
206°C to 210°C | |
C6H15Cl2N | |
MFCD00012519 | |
3591991 | |
RAGSWDIQBBZLLL-UHFFFAOYSA-N | |
2-chloro-N,N-diethylethanamine;hydrochloride | |
13363 | |
≥98% |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318-H341
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : YE1050000
missing translation for 'tsca' : TSCA
RUO – Research Use Only