missing translation for 'onlineSavingsMsg'
Learn More

2-(Diisopropylphosphino)ethylamine, 10% w/w soln. in THF

Product Code. 15485746
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15485746

Brand: Thermo Scientific Alfa Aesar H60435.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1053657-14-9 | C8H20NP | 161.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1053657-14-9
Molecular Formula C8H20NP
Molecular Weight (g/mol) 161.23
MDL Number MFCD11044863
InChI Key QUVUVMRUSAKOPB-UHFFFAOYSA-N
Synonym 2-diisopropylphosphino ethylamine, 2-di-i-propylphosphino ethylamine, 2-bis 1-methylethyl phosphino ethanamine, acmc-20aonc, 2-aminoethyldiisopropylphosphine, 2-di-isopropylphosphino ethanamine, 2-aminoethyl diisopropylphosphane, 2-di propan-2-yl phosphanyl ethan-1-amine, 2-aminoethyl bis propan-2-yl phosphane, 2-diisopropylphosphino ethylamine, kanata purity
PubChem CID 57552996
IUPAC Name 2-di(propan-2-yl)phosphanylethanamine
SMILES CC(C)P(CCN)C(C)C

Specifications

Linear Formula [(CH3)2CH]2PCH2CH2NH2
Quantity 5 g
UN Number UN2924
Sensitivity Air Sensitive
Formula Weight 161.22
Concentration or Composition (by Analyte or Components) 10% w/w soln. in THF
Chemical Name or Material 2-(Diisopropylphosphino)ethylamine
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-(Diisopropylphosphino)ethylamine, 10% w/w solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, 2-(Diisopropylphosphino)ethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.