missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)ethyl methacrylate, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10666992
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2867-47-2
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00008589
InChI Key JKNCOURZONDCGV-UHFFFAOYSA-N
Synonym 2-dimethylamino ethyl methacrylate, dimethylaminoethyl methacrylate, ageflex fm-1, dmaema, usaf rh-3, 2-propenoic acid, 2-methyl-, 2-dimethylamino ethyl ester, 2-n,n-dimethylamino ethyl methacrylate, n,n-dimethylaminoethyl methacrylate, 2-dimethylaminoethyl methacrylate, n,n-dimethylethanolamine methacrylate
PubChem CID 17869
IUPAC Name 2-(dimethylamino)ethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCN(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless to Yellow
Density 0.9330g/mL
Boiling Point 182.0°C to 192.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(CH3)CO2CH2CH2N(CH3)2
Refractive Index 1.4390 to 1.4410
Quantity 250 g
Beilstein 04, III, 649
Specific Gravity 0.933
Solubility Information Solubility in water: soluble.
Stabilizer 750 to 2000ppm MEHQ
Viscosity 1.32 mPa.s (20°C)
Formula Weight 157.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Dimethylamino)ethyl methacrylate, Stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'