missing translation for 'onlineSavingsMsg'
Learn More

2-(Dimethylamino)thiazole-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11996781
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1005-28-3
InChI Key ZOPJEHPVWLTDJW-UHFFFAOYSA-N
Synonym 2-dimethylamino-1,3-thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carbaldehyde, 2-dimethylamino thiazole-5-carboxaldehyde, 2-dimethylamino-thiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde,2-dimethylamino, 5-thiazolecarboxaldehyde, 2-dimethylamino, 2-dimethylaminothiazole-5-carbaldehyde
PubChem CID 12846795
IUPAC Name 2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
SMILES CN(C)C1=NC=C(S1)C=O

Specifications

Melting Point 75°C to 77°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H8N2OS
Quantity 1 g
Molecular Weight (g/mol) 156.21
Formula Weight 156.21
Percent Purity 97%
Chemical Name or Material 2-(Dimethylamino)thiazole-5-carboxaldehyde
compliance-icons
Product Identifier
  • 2-(Dimethylamino)thiazole-5-carboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.