Learn More
2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 13242-44-9 |
---|---|
Molecular Formula | C4H11NS·HCl |
Molecular Weight (g/mol) | 141.67 |
MDL Number | MFCD00012633 |
InChI Key | NRVFDGZJTPCULU-UHFFFAOYSA-N |
Synonym | 2-dimethylamino ethanethiol hydrochloride, captamine hydrochloride, meda, captamine hcl, n,n-dimethylcysteamine hydrochloride, dimethylaminoethanethiol hydrochloride, 2-dimethylaminoethanethiol hydrochloride, n-dimethylcysteamine hydrochloride, 2-mercaptoethyl dimethylammonium chloride, unii-m43ax41u87 |
PubChem CID | 25798 |
IUPAC Name | 2-(dimethylamino)ethanethiol;hydrochloride |
SMILES | CN(C)CCS.Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13242-44-9 | |
141.67 | |
NRVFDGZJTPCULU-UHFFFAOYSA-N | |
25798 | |
CN(C)CCS.Cl |
C4H11NS·HCl | |
MFCD00012633 | |
2-dimethylamino ethanethiol hydrochloride, captamine hydrochloride, meda, captamine hcl, n,n-dimethylcysteamine hydrochloride, dimethylaminoethanethiol hydrochloride, 2-dimethylaminoethanethiol hydrochloride, n-dimethylcysteamine hydrochloride, 2-mercaptoethyl dimethylammonium chloride, unii-m43ax41u87 | |
2-(dimethylamino)ethanethiol;hydrochloride |
Specifications
13242-44-9 | |
100.0 | |
94% min. (Iodimetry) | |
C4H11NS·HCl | |
MFCD00012633 | |
Solubility in water: soluble in water | |
CN(C)CCS.Cl | |
141.67 | |
141.67 | |
2-Dimethylaminoethanethiol hydrochloride, 95% |
94.0 | |
154°C to 159°C | |
Glass bottle | |
(CH3)2NCH2CH2SH·HCl | |
2-dimethylamino ethanethiol hydrochloride, captamine hydrochloride, meda, captamine hcl, n,n-dimethylcysteamine hydrochloride, dimethylaminoethanethiol hydrochloride, 2-dimethylaminoethanethiol hydrochloride, n-dimethylcysteamine hydrochloride, 2-mercaptoethyl dimethylammonium chloride, unii-m43ax41u87 | |
NRVFDGZJTPCULU-UHFFFAOYSA-N | |
2-(dimethylamino)ethanethiol;hydrochloride | |
25798 | |
95% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 236-221-6
missing translation for 'rtecsNumber' : KJ1051000
missing translation for 'tsca' : TSCA
- 2-Dimethylaminoethanethiol hydrochloride
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.