missing translation for 'onlineSavingsMsg'
Learn More
2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals Product Code.: 15471396

2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15471396
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15471396

Brand: Thermo Scientific Alfa Aesar H54614.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1006876-27-2
Molecular Formula C14H21BN2O3
Molecular Weight (g/mol) 276.143
MDL Number MFCD08458484
InChI Key RNCGUUHRBRONRG-UHFFFAOYSA-N
Synonym n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl picolinamide, 6-dimethylcarbamoyl pyridin-3-ylboronic acid pinacol ester, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 6-dimethylcarbamoyl pyridine-3-boronic acid pinacol ester, 2-dimethylcarbamoyl pyridine-5-boronic acid pinacol ester, n,n-dimethyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-2-carboxamide, 2-n,n-dimthylaminocarbonyl pyridine-5-boronic acid pinacol ester, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl picolinamide, 2-pyridinecarboxamide, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n,n-dimethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinecarboxamide
PubChem CID 57416491
IUPAC Name N,N-dimethyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine-2-carboxamide
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)C(=O)N(C)C

Specifications

Quantity 250 mg
Formula Weight 276.14
Percent Purity 96%
Chemical Name or Material 2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberL07X021 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54614.MD,H54614.03
Lot NumberZ13F019 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54614.MD,H54614.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Dimethylcarbamoyl)pyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.