missing translation for 'onlineSavingsMsg'
Learn More

2-(Diphenylphosphino)ethylammonium tetrafluoroborate, 97+%, Thermo Scientific™

Product Code. 15415676
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1222630-32-1
Molecular Formula C14H17BF4NP
Molecular Weight (g/mol) 317.07
MDL Number MFCD17014009
InChI Key ACXZLRCRPLYXBQ-UHFFFAOYSA-O
Synonym 2-diphenylphosphino ethanaminium tetrafluoroborate, 2-ammonioethyl diphenylphosphane tetrafluoroborate, 2-diphenylphosphanyl ethan-1-aminium tetrafluoroborate, 2-diphenylphosphino ethylammonium tetrafluoroborate
PubChem CID 71463643
IUPAC Name 2-diphenylphosphanylethylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.[NH3+]CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 116°C to 122°C
Linear Formula (C6H5)2PCH2CH2NH3·BF4
Quantity 5 g
UN Number UN3263
Formula Weight 317.06
Percent Purity ≥97%
Chemical Name or Material 2-(Diphenylphosphino)ethylammonium tetrafluoroborate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'