missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyacetyl chloride, 97%, stab. with 0.3% Magnesium oxide, Thermo Scientific Chemicals

70.75 € - 630.14 €

Chemical Identifiers

CAS 14077-58-8
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD11170566
InChI Key ZPMWWAIBJJFPPQ-UHFFFAOYSA-N
PubChem CID 84189
IUPAC Name 2-ethoxyacetyl chloride
SMILES CCOCC(=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15468945
View Documents
Thermo Scientific Alfa Aesar
H34443.06
5 g
70.75 €
5g
Please to purchase this item. Need a web account? Register with us today!
15478945
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
H34443.14
25 g
198.99 €
25g
Please to purchase this item. Need a web account? Register with us today!
15488945
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
H34443.22
100 g
630.14 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in fine chemical syntheses and used as pharmaceutical intermediates. A trace of cis-ethoxyacetamide was also detected and isolated with an authentic sample prepared from the reaction of cis-3-amino-6-methylchroman-4-01 (2) with 2-ethoxyacetyl chloride.

Solubility
Insoluble in water.

Notes
Reacts with strong oxidizing agents. Store at room temperature.
Specifications

Chemical Identifiers

14077-58-8
122.548
ZPMWWAIBJJFPPQ-UHFFFAOYSA-N
2-ethoxyacetyl chloride
C4H7ClO2
MFCD11170566
84189
CCOCC(=O)Cl

Specifications

14077-58-8
CH3CH2OCH2COCl
UN2920
Insoluble in water.
CCOCC(=O)Cl
122.548
122.55
2-Ethoxyacetyl chloride, stabilized with 0.3% Magnesium oxide
C4H7ClO2
MFCD11170566
ethoxyacetyl chloride, ethoxyacetylchloride, ethoxy-acetyl chloride, acetyl chloride,2-ethoxy, 2-ethoxyacetylchloride, ethoxy acetyl chloride, ethoxyacetic acid chloride, 2-ethoxy-acetyl chloride, acetyl chloride, 2-ethoxy
ZPMWWAIBJJFPPQ-UHFFFAOYSA-N
2-ethoxyacetyl chloride
84189
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H226-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H226-H314-H332-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

missing translation for 'einecsNumber' : 237-930-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only