Learn More
Alfa Aesar™ 2-Ethoxybenzoyl chloride, 98%
Chemical Identifiers
CAS | 42926-52-3 |
---|---|
Molecular Formula | C9H9ClO2 |
Molecular Weight (g/mol) | 184.619 |
MDL Number | MFCD00016348 |
InChI Key | MDKAAWDKKBFSTK-UHFFFAOYSA-N |
PubChem CID | 3016397 |
IUPAC Name | 2-ethoxybenzoyl chloride |
SMILES | CCOC1=CC=CC=C1C(=O)Cl |
Chemical Identifiers
42926-52-3 | |
184.619 | |
MDKAAWDKKBFSTK-UHFFFAOYSA-N | |
2-ethoxybenzoyl chloride |
C9H9ClO2 | |
MFCD00016348 | |
3016397 | |
CCOC1=CC=CC=C1C(=O)Cl |
Specifications
42926-52-3 | |
C9H9ClO2 | |
UN3265 | |
MDKAAWDKKBFSTK-UHFFFAOYSA-N | |
2-ethoxybenzoyl chloride | |
3016397 | |
98% |
114°C to 118°C (5 mmHg) | |
MFCD00016348 | |
o-ethoxybenzoyl chloride, benzoyl chloride, 2-ethoxy, 2-ethoxybenzoylchloride, acmc-209jrt, 2-ethoxy-benzoyl chloride, benzoyl chloride,2-ethoxy, 2-ethoxybenzoic acid chloride, ksc491c5l, 2-ethoxybenzoyl chloride | |
CCOC1=CC=CC=C1C(=O)Cl | |
184.619 | |
184.62 | |
2-Ethoxybenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.