missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxycyclohexanone, 98%, Thermo Scientific Chemicals

17.30 € - 164.00 €

Chemical Identifiers

CAS 33371-97-0
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00015377
InChI Key WBPWDGRYHFQTRC-QMMMGPOBSA-N
Synonym 2-ethoxycyclohexanone, ethoxycyclohexanone, 2-ethoxy-cyclohexanone, acmc-1ai48
PubChem CID 2733330
SMILES CCO[C@H]1CCCCC1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11406550
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
H27613.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11416550
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
H27613.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11426550
missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
H27613.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33371-97-0
142.20
WBPWDGRYHFQTRC-QMMMGPOBSA-N
2733330
C8H14O2
MFCD00015377
2-ethoxycyclohexanone, ethoxycyclohexanone, 2-ethoxy-cyclohexanone, acmc-1ai48
CCO[C@H]1CCCCC1=O

Specifications

33371-97-0
85°C (15 mmHg)
C8H14O2
MFCD00015377
2-ethoxycyclohexanone, ethoxycyclohexanone, 2-ethoxy-cyclohexanone, acmc-1ai48
CCO[C@H]1CCCCC1=O
142.20
142.2
2-Ethoxycyclohexanone
1.002
54°C (129°F)
1.451
UN1224
WBPWDGRYHFQTRC-QMMMGPOBSA-N
(2S)-2-ethoxycyclohexan-1-one
2733330
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.