Learn More
2-Ethyl-1-hexanol, 99%, Thermo Scientific Chemicals
19.20 € - 347.73 €
Chemical Identifiers
CAS | 104-76-7 |
---|---|
MDL Number | MFCD00004746 |
InChI Key | YIWUKEYIRIRTPP-UHFFFAOYSA-N |
Synonym | 2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151 |
PubChem CID | 7720 |
ChEBI | CHEBI:16011 |
IUPAC Name | 2-ethylhexan-1-ol |
SMILES | CCCCC(CC)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10773851
|
Thermo Scientific Acros
118530050 |
5 mL | Glass Bottle | N/A | |||||
10102580
|
Thermo Scientific Acros
118530010 |
1 L | Glass Bottle |
38.49 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10488450
|
Thermo Scientific Acros
118530025 |
2.5 L | Glass Bottle |
64.12 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10063803
|
Thermo Scientific Acros
118530250 |
25 L | Metal drum |
347.73 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-76-7 | |
YIWUKEYIRIRTPP-UHFFFAOYSA-N | |
7720 | |
2-ethylhexan-1-ol |
MFCD00004746 | |
2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151 | |
CHEBI:16011 | |
CCCCC(CC)CO |
Specifications
104-76-7 | |
100.0 | |
Authentic | |
C8H18O | |
1.4300 to 1.4330 (20°C, 589nm) | |
15, 3863 | |
YIWUKEYIRIRTPP-UHFFFAOYSA-N | |
2-ethylhexan-1-ol | |
7720 | |
130.23 | |
Liquid |
98.5 | |
0.8330g/mL | |
99% | |
CH3(CH2)3CH(C2H5)CH2OH | |
MFCD00004746 | |
2-ethylhexanol, 2-ethyl-1-hexanol, 1-hexanol, 2-ethyl, 2-ethylhexyl alcohol, ethylhexanol, alcohol, 2-ethylhexyl, xi-2-ethyl-1-hexanol, 2-aethylhexanol german, ethylhexanol, 2, fema no. 3151 | |
CCCCC(CC)CO | |
130.231 | |
CHEBI:16011 | |
≥98.5% | |
2-Ethyl-1-hexanol |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only