missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-oxazoline, 99%, Thermo Scientific Chemicals

Product Code. 11410897
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-ethyl-2-oxazoline is an organic intermediate and monomer that is used in water-soluble polymers, pharmaceuticals, cosmetics, specialty coatings and adhesives.

Solubility
Fully miscible in water.

Notes
TRUSTED_SUSTAINABILITY

Specifications

CAS 10431-98-8
Melting Point -62°C
Density 0.981
pH 10 to 12
Boiling Point 128°C to 129°C
Flash Point 29°C (84°F)
Odor Amine-like
Molecular Formula C5H9NO
Refractive Index 1.437
MDL Number MFCD00005302
Quantity 100 g
UN Number UN2734
Beilstein 105464
Sensitivity Air Sensitive
Synonym 2-ethyl-2-oxazoline, 2-ethyloxazoline, 2-ethyl-4,5-dihydrooxazole, oxazole, 2-ethyl-4,5-dihydro, 2-oxazoline, 2-ethyl, unii-b8cd92t4p5, unii-jm7uh9q3yn, unii-hnx7574gtx, unii-s5dpg6k67i, ethyl oxazoline
Solubility Information Fully miscible in water.
InChI Key NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES CCC1=NCCO1
IUPAC Name 2-ethyl-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol) 99.13
PubChem CID 66412
Formula Weight 99.13
Percent Purity 99%
Chemical Name or Material 2-Ethyl-2-oxazoline
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'