missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-6-methylphenol, 97%, Thermo Scientific Chemicals

33.47 € - 473.36 €

Chemical Identifiers

CAS 1687-64-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00962292
InChI Key CIRRFAQIWQFQSS-UHFFFAOYSA-N
PubChem CID 519333
IUPAC Name 2-ethyl-6-methylphenol
SMILES CCC1=CC=CC(=C1O)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11460708
View Documents
Thermo Scientific Alfa Aesar
B25213.03
1 g
33.47 €
1g
Please to purchase this item. Need a web account? Register with us today!
11470708
View Documents
Thermo Scientific Alfa Aesar
B25213.06
5 g
122.61 €
5g
Please to purchase this item. Need a web account? Register with us today!
11480708
View Documents
Thermo Scientific Alfa Aesar
B25213.14
25 g
473.36 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-6-methylphenol is used as pharmaceutical intermediate.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications

Chemical Identifiers

1687-64-5
136.194
CIRRFAQIWQFQSS-UHFFFAOYSA-N
2-ethyl-6-methylphenol
C9H12O
MFCD00962292
519333
CCC1=CC=CC(=C1O)C

Specifications

1687-64-5
1.531
phenol, 2-ethyl-6-methyl, o-cresol, 6-ethyl, 6-ethyl-o-cresol, unii-359wuc62ri, 2-ethyl-6-methyl-phenol, 6-ethyl-2-methylphenol, acmc-20anwj, 2-methyl-6-ethylphenol, 2-ethyl-6-methyl phenol, phenol,2-ethyl-6-methyl
CCC1=CC=CC(=C1O)C
136.194
136.19
2-Ethyl-6-methylphenol
C9H12O
MFCD00962292
CIRRFAQIWQFQSS-UHFFFAOYSA-N
2-ethyl-6-methylphenol
519333
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only