Learn More
2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals
82.71 € - 228.14 €
Chemical Identifiers
CAS | 2736-40-5 |
---|---|
Molecular Formula | C6H11ClO |
Molecular Weight (g/mol) | 134.60 |
MDL Number | MFCD00018812 |
InChI Key | SMUKODJVMQOSAB-UHFFFAOYSA-N |
Synonym | 2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride |
PubChem CID | 75954 |
IUPAC Name | 2-ethylbutanoyl chloride |
SMILES | CCC(CC)C(Cl)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11430228
|
Thermo Scientific Alfa Aesar
B23774.14 |
25 g |
82.71 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440228
|
Thermo Scientific Alfa Aesar
B23774.22 |
100 g |
228.14 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylbutyryl chloride was used in syntheses of bicyclic thiophene derivatives. It is used as an organic intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store under inert gas. Store away from oxidizing agents, amines, alcohols, active metals, water/ moisture.
Chemical Identifiers
2736-40-5 | |
134.60 | |
SMUKODJVMQOSAB-UHFFFAOYSA-N | |
75954 | |
CCC(CC)C(Cl)=O |
C6H11ClO | |
MFCD00018812 | |
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride | |
2-ethylbutanoyl chloride |
Specifications
2736-40-5 | |
137°C to 139°C | |
Acrid | |
1.425 | |
UN3094 | |
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride | |
SMUKODJVMQOSAB-UHFFFAOYSA-N | |
2-ethylbutanoyl chloride | |
75954 | |
99% |
0.98 | |
33°C (91°F) | |
C6H11ClO | |
MFCD00018812 | |
Moisture sensitive | |
Reacts with water. | |
CCC(CC)C(Cl)=O | |
134.60 | |
134.61 | |
2-Ethylbutyryl chloride |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 220-359-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.