missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanal, 96%, Thermo Scientific Chemicals

32.86 € - 81.61 €

Chemical Identifiers

CAS 123-05-7
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00006987
InChI Key LGYNIFWIKSEESD-UHFFFAOYSA-N
Synonym hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde
PubChem CID 31241
IUPAC Name 2-ethylhexanal
SMILES CCCCC(CC)C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11445467
View Documents
Thermo Scientific Alfa Aesar
B23471.18
50 g
32.86 €
50g
Please to purchase this item. Need a web account? Register with us today!
11455467
View Documents
Thermo Scientific Alfa Aesar
B23471.30
250 g
81.61 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-Ethylhexanal is used as an intermediate for chemical synthesis and in the production of pharmaceuticals and odorous substances.  

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanal is used as an intermediate for chemical synthesis & in the production of disinfectants, pharmaceuticals and odorous substances.  

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Specifications

Chemical Identifiers

123-05-7
128.215
LGYNIFWIKSEESD-UHFFFAOYSA-N
31241
CCCCC(CC)C=O
C8H16O
MFCD00006987
hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde
2-ethylhexanal

Specifications

123-05-7
0.819
50°C (122°F)
1.4155
UN1989
Air Sensitive
Slightly soluble in water.
CCCCC(CC)C=O
128.215
128.21
2-Ethylhexanal
<-100°C
163°C
C8H16O
MFCD00006987
1700556
hexanal, 2-ethyl, 2-ethylhexaldehyde, 2-ethylhexylaldehyde, 3-formylheptane, 2-ethylcaproaldehyde, butylethylacetaldehyde, ethylbutylacetaldehyde, ethylhexaldehyde, alpha-ethylcaproaldehyde, butyl ethyl acetaldehyde
LGYNIFWIKSEESD-UHFFFAOYSA-N
2-ethylhexanal
31241
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P210-P233-P240-P241-P242-P243-P261-P272-P280g-P281-P303+P361+P353-P308+P313-P333+P313-P363-P370+P378q-P501c

H226-H317-H361

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

missing translation for 'einecsNumber' : 204-596-5

missing translation for 'rtecsNumber' : MN7525000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Ethylhexanal, 96%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.