missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

Product Code. 11329629
Click to view available options
Quantity:
10,000 mL
100 mL
2500 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

Solubility
Miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-57-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00002675
InChI Key OBETXYAYXDNJHR-UHFFFAOYSA-N
Synonym 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
PubChem CID 8697
IUPAC Name 2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O

Specifications

Melting Point -59°C
Density 0.91
pH 3
Boiling Point 226°C to 229°C
Flash Point 114°C (237°F)
Refractive Index 1.425
Quantity 100 mL
Beilstein 1750468
Solubility Information Miscible with water.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 2-Ethylhexanoic acid
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'