Learn More
2-Ethylhexylamine, 98%, Thermo Scientific Chemicals
24.92 € - 164.82 €
Chemical Identifiers
CAS | 104-75-6 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.247 |
MDL Number | MFCD00008148 |
InChI Key | LTHNHFOGQMKPOV-UHFFFAOYSA-N |
Synonym | 2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame |
PubChem CID | 7719 |
IUPAC Name | 2-ethylhexan-1-amine |
SMILES | CCCCC(CC)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494183
|
Thermo Scientific Alfa Aesar
A10372.AE |
100 mL |
24.92 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404193
|
Thermo Scientific Alfa Aesar
A10372.AP |
500 mL | N/A | ||||||
11414193
|
Thermo Scientific Alfa Aesar
A10372.0F |
2500 mL |
164.82 €
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
Solubility
Soluble in water. 2.5 g/L (20°C)
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Protect against electrostatic charges. Store away from acid chlorides and acid anhydrides.
Chemical Identifiers
104-75-6 | |
129.247 | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
7719 | |
CCCCC(CC)CN |
C8H19N | |
MFCD00008148 | |
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame | |
2-ethylhexan-1-amine |
Specifications
104-75-6 | |
0.789 | |
50°C (122°F) | |
C8H19N | |
MFCD00008148 | |
1209249 | |
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
2-ethylhexan-1-amine | |
7719 | |
98% |
-76°C | |
169°C | |
Amine-like | |
1.43 | |
UN2276 | |
Air Sensitive | |
Soluble in water. 2.5g/L (20°C) | |
CCCCC(CC)CN | |
129.247 | |
129.25 | |
2-Ethylhexylamine |
Safety and Handling
GHS H Statement
H311-H314-H318-H226-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P302+P352-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P330-P362-P370+P378q-P501c
H226-H302-H312-H314-H318-H331
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 2-ETHYLHEXYLAMINE
missing translation for 'einecsNumber' : 203-233-8
missing translation for 'rtecsNumber' : MQ5250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only