missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10722133
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 365-12-8
Molecular Formula C13H8FO2
Molecular Weight (g/mol) 215.20
MDL Number MFCD00278732
InChI Key SLKZDWAZOKIEEU-UHFFFAOYSA-M
Synonym 2'-fluorobiphenyl-4-carboxylic acid, 4-2-fluorophenyl benzoic acid, 2'-fluoro-1,1'-biphenyl-4-carboxylic acid, 2'-fluoro 1,1'-biphenyl-4-carboxylic acid, 2'-fluoro-biphenyl-4-carboxylic acid, maybridge3_006504, amtda094, acmc-1cr23, 4-2-fluorophenyl-benzoic acid, 2'-fluoro-4-biphenylcarboxylic acid
PubChem CID 715022
SMILES [O-]C(=O)C1=CC=C(C=C1)C1=CC=CC=C1F

Specifications

Assay Percent Range 95%
Linear Formula FC6H4C6H4CO2H
Quantity 250 mg
IUPAC Name 2'-fluoro-[1,1'-biphenyl]-4-carboxylate
Formula Weight 216.21
Percent Purity 95%
Chemical Name or Material 2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'