missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitrobenzoic acid, 97%

Product Code. 15449828
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15449828

Brand: Thermo Scientific Alfa Aesar H63235.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 317-46-4 | C7H4FNO4 | 185.11 g/mol

2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-nitrobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 317-46-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD06797825
InChI Key WLGUSLGYTNJJFV-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitrobenzoic acid, 2-fluoro-3-nitrobenzoicacid, 2-fluoro-3-nitro benzoic acid, 3-carboxy-2-fluoronitrobenzene, pubchem1398, acmc-1ajf3, ksc494k8n, benzoic acid,2-fluoro-3-nitro
PubChem CID 18672992
IUPAC Name 2-fluoro-3-nitrobenzoic acid
SMILES OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O

Specifications

Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 185.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.