missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-(trifluoromethoxy)-DL-phenylalanine, 97%, Thermo Scientific™
70.00 € - 230.00 €
Chemical Identifiers
CAS | 1391008-15-3 |
---|---|
Molecular Formula | C10H9F4NO3 |
Molecular Weight (g/mol) | 267.18 |
MDL Number | MFCD16652500 |
InChI Key | POFOCSWADMUTGH-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-trifluoromethoxy-dl-phenylalanine, 2-amino-3-2-fluoro-4-trifluoromethoxy phenyl propanoic acid |
PubChem CID | 66523488 |
IUPAC Name | 2-amino-3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid |
SMILES | C1=CC(=C(C=C1OC(F)(F)F)F)CC(C(=O)O)N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1391008-15-3 | |
267.18 | |
POFOCSWADMUTGH-UHFFFAOYSA-N | |
66523488 | |
C1=CC(=C(C=C1OC(F)(F)F)F)CC(C(=O)O)N |
C10H9F4NO3 | |
MFCD16652500 | |
2-fluoro-4-trifluoromethoxy-dl-phenylalanine, 2-amino-3-2-fluoro-4-trifluoromethoxy phenyl propanoic acid | |
2-amino-3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid |
Specifications
1391008-15-3 | |
MFCD16652500 | |
POFOCSWADMUTGH-UHFFFAOYSA-N | |
2-amino-3-[2-fluoro-4-(trifluoromethoxy)phenyl]propanoic acid | |
66523488 | |
97% |
C10H9F4NO3 | |
2-fluoro-4-trifluoromethoxy-dl-phenylalanine, 2-amino-3-2-fluoro-4-trifluoromethoxy phenyl propanoic acid | |
C1=CC(=C(C=C1OC(F)(F)F)F)CC(C(=O)O)N | |
267.18 | |
267.18 | |
2-Fluoro-4-(trifluoromethoxy)-DL-phenylalanine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures