missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxybenzoic acid, 97+%, Thermo Scientific Chemicals

Product Code. 15429047
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-methoxybenzoic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-83-9
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD00272566
InChI Key REDSLTKMNCCQBC-UHFFFAOYSA-N
PubChem CID 2774543
IUPAC Name 2-fluoro-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)F)C(=O)O

Specifications

Quantity 250 mg
Synonym 2-fluoro-5-methoxy-benzoic acid, benzoic acid,2-fluoro-5-methoxy, benzoic acid, 2-fluoro-5-methoxy, pubchem4966, 6-fluoro-m-anisic acid, 3-carboxy-4-fluoroanisole, 2-fluoro-5-methoxybenzoicacid, 6-fluoro-3-methoxybenzoic acid, rarechem al bo 0748, buttpark 45\01-62
Formula Weight 170.14
Percent Purity ≥97%
Chemical Name or Material 2-Fluoro-5-methoxybenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'