missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitroaniline, 98%, Thermo Scientific Chemicals

Product Code. 10377870
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 369-36-8
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007652
InChI Key KJVBJICWGQIMOZ-UHFFFAOYSA-N
Synonym benzenamine, 2-fluoro-5-nitro, 2-fluoro-5-nitrophenylamine, 2-fluoro-5-nitro-phenylamine, 1-amino-2-fluoro-5-nitrobenzene, 2-fluoro-5-nitrobenzenamine, 2-fluoro-5-nitro aniline, 3-amino-4-fluoronitrobenzene, pubchem8504, acmc-1crtl, 5-nitro-2-fluoroaniline
PubChem CID 67785
IUPAC Name 2-fluoro-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 99°C to 103°C
Color Brown to Ochre-Yellow
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(NO2)NH2
Quantity 100 g
Beilstein 12, I, 355
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,alcohol and toluene
Formula Weight 156.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-nitroaniline, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluoro-5-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Flammable solid Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.