missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11394096
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Fluoro-5-nitrobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-nitrobenzaldehyde, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27996-87-8
Molecular Formula C7H4FNO3
Molecular Weight (g/mol) 169.111
MDL Number MFCD00042298
InChI Key VVXFDFQEIRGULC-UHFFFAOYSA-N
Synonym fluoro nitrobenzaldehyde, 5-nitro-2-fluorobenzaldehyde, 4-fluoro-3-formylnitrobenzene, 2-fluoro-5-nitro-benzaldehyde, benzaldehyde, 2-fluoro-5-nitro, pubchem1459, acmc-209h0h, akos bb460, asischem d51510, 2-fluoro-5 nitrobenzaldehyde
PubChem CID 2734770
IUPAC Name 2-fluoro-5-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C=O)F

Specifications

Melting Point 57°C to 59°C
Quantity 5 g
Beilstein 1950329
Sensitivity Air Sensitive
Solubility Information It is insoluble in water.
Formula Weight 169.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-nitrobenzaldehyde

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'