Learn More
2-Fluoro-5-nitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals
63.10 € - 251.00 €
Chemical Identifiers
CAS | 819849-20-2 |
---|---|
Molecular Formula | C6H5BFNO4 |
Molecular Weight (g/mol) | 184.92 |
MDL Number | MFCD03092929 |
InChI Key | SYAFEPQKPQIRAL-UHFFFAOYSA-N |
Synonym | 2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 2-fluoro-5-nitrobenzeneboronic acid, 5-nitro-2-fluorophenylboronic acid, boronic acid, 2-fluoro-5-nitrophenyl, 2-fluoro-5-nitro-phenyl boronic acid, acmc-209pmq, ksc649e7n, 2-fluoro-5-nitro phenyl boronic acid, b-2-fluoro-5-nitrophenyl-boronic acid |
PubChem CID | 2763280 |
IUPAC Name | (2-fluoro-5-nitrophenyl)boronic acid |
SMILES | OB(O)C1=CC(=CC=C1F)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15491139
|
Thermo Scientific Alfa Aesar
H64100.03 |
1 g |
63.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15401149
|
Thermo Scientific Alfa Aesar
H64100.06 |
5 g |
251.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Fluoro-5-nitrobenzeneboronic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluoro-5-nitrobenzeneboronic acid is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
819849-20-2 | |
184.92 | |
SYAFEPQKPQIRAL-UHFFFAOYSA-N | |
2763280 | |
OB(O)C1=CC(=CC=C1F)[N+]([O-])=O |
C6H5BFNO4 | |
MFCD03092929 | |
2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 2-fluoro-5-nitrobenzeneboronic acid, 5-nitro-2-fluorophenylboronic acid, boronic acid, 2-fluoro-5-nitrophenyl, 2-fluoro-5-nitro-phenyl boronic acid, acmc-209pmq, ksc649e7n, 2-fluoro-5-nitro phenyl boronic acid, b-2-fluoro-5-nitrophenyl-boronic acid | |
(2-fluoro-5-nitrophenyl)boronic acid |
Specifications
819849-20-2 | |
C6H5BFNO4 | |
2-fluoro-5-nitrophenyl boronic acid, 2-fluoro-5-nitrophenylboronicacid, 2-fluoro-5-nitrobenzeneboronic acid, 5-nitro-2-fluorophenylboronic acid, boronic acid, 2-fluoro-5-nitrophenyl, 2-fluoro-5-nitro-phenyl boronic acid, acmc-209pmq, ksc649e7n, 2-fluoro-5-nitro phenyl boronic acid, b-2-fluoro-5-nitrophenyl-boronic acid | |
SYAFEPQKPQIRAL-UHFFFAOYSA-N | |
(2-fluoro-5-nitrophenyl)boronic acid | |
2763280 | |
97% | |
2-Fluoro-5-nitrobenzeneboronic acid |
White | |
MFCD03092929 | |
Slightly soluble in water. | |
OB(O)C1=CC(=CC=C1F)[N+]([O-])=O | |
184.92 | |
184.92 | |
Powder |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.