missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrotoluene, 98+%, Thermo Scientific Chemicals

Product Code. 11487606
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 455-88-9
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00007284
InChI Key XUCYJGMIICONES-UHFFFAOYSA-N
Synonym 2-fluoro-5-nitrotoluene, benzene, 1-fluoro-2-methyl-4-nitro, 4-fluoro-3-methylnitrobenzene, toluene, 2-fluoro-5-nitro, 1-fluoro-2-methyl-4-nitro-benzene, 5-nitro-2-fluorotoluene, 3-methyl-4-fluoronitrobenzene, pubchem1593, 2-fluoro-5-nitro-toluene, 5-nitro-2-fluoro toluene
PubChem CID 68001
IUPAC Name 1-fluoro-2-methyl-4-nitrobenzene
SMILES CC1=CC(=CC=C1F)[N+]([O-])=O

Specifications

Melting Point 37°C to 40°C
Boiling Point 98°C to 101°C (13 mmHg)
Flash Point 105°C (221°F)
Quantity 10 g
UN Number UN2811
Beilstein 1940341
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 2-Fluoro-5-nitrotoluene
compliance-icons
Product Identifier
  • 2-Fluoro-5-nitrotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-fluoro-2-methyl-4-nitro-Benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'