missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoro-6'-hydroxyacetophenone, 98%, Thermo Scientific™

Product Code. 11322129
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93339-98-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD01090995
InChI Key PSNPXFMLAVLPPP-UHFFFAOYSA-N
PubChem CID 2737324
IUPAC Name 1-(2-fluoro-6-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(O)C=CC=C1F

Specifications

Refractive Index 1.534
Quantity 1 g
Synonym 1-2-fluoro-6-hydroxyphenyl ethanone, 2'-fluoro-6'-hydroxyacetophenone, 2-fluoro-6-hydroxyacetophenone, 1-2-fluoro-6-hydroxyphenyl ethan-1-one, 1-2-fluoro-6-hydroxy-phenyl-ethanone, ethanone, 1-2-fluoro-6-hydroxyphenyl, 1-acetyl-2-fluoro-6-hydroxybenzene, 1-2-fluoranyl-6-oxidanyl-phenyl ethanone, ethanone,1-2-fluoro-6-hydroxyphenyl
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 2'-Fluoro-6'-hydroxyacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'