missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals Product Code.: 15417876

2-Fluoro-6-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Product Code. 15417876
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15417876

Brand: Thermo Scientific Alfa Aesar H62812.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1599432-41-3
Molecular Formula C13H18BFO3
Molecular Weight (g/mol) 252.092
InChI Key CSXSULPDIWBLHT-UHFFFAOYSA-N
Synonym 2-2-fluoro-6-methoxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-fluoro-6-methoxybenzeneboronic acid pinacol ester, 2-fluoro-6-methoxyphenylboronic acid pinacol ester, 2-fluoro-6-methoxybenzeneboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronicacidpinacolester, 2-fluoro-6-methoxyphenylboronic acid 2,3-dimethyl-2,3-butanediyl
PubChem CID 72221147
IUPAC Name 2-(2-fluoro-6-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CC=C2F)OC

Specifications

Quantity 250 mg
Formula Weight 252.09
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-6-methoxybenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoro-6-methoxybenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.