missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-phenoxybenzonitrile, 95%, Thermo Scientific™

Product Code. 11499100
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11499100

Brand: Thermo Scientific Alfa Aesar A14566.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175204-06-5
Molecular Formula C13H8FNO
Molecular Weight (g/mol) 213.211
MDL Number MFCD00068201
InChI Key FBHXELGUZCMKQY-UHFFFAOYSA-N
Synonym 2-fluoro-6-phenyloxybenzonitrile, 2-cyano-3-fluorodiphenyl ether, 6-fluoro-2-phenoxybenzenecarbonitrile, acmc-1bwys, maybridge1_005593, 2-fluoro-6-phenoxybenzonitrile, 2-fluoro-6-phenoxy benzonitrile, 2-fluoranyl-6-phenoxy-benzenecarbonitrile, 2-fluoro-6-phenoxybenzonitrile, tech
PubChem CID 2737453
IUPAC Name 2-fluoro-6-phenoxybenzonitrile
SMILES C1=CC=C(C=C1)OC2=C(C(=CC=C2)F)C#N

Specifications

Melting Point 90°C to 93°C
Quantity 5 g
Formula Weight 213.21
Percent Purity 95%
Chemical Name or Material 2-Fluoro-6-phenoxybenzonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato