missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzothiazole, 99%, Thermo Scientific Chemicals

Product Code. 11394226
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1123-98-4
Molecular Formula C7H4FNS
Molecular Weight (g/mol) 153.174
MDL Number MFCD00047254
InChI Key QVWCHVAUHZEAAT-UHFFFAOYSA-N
Synonym 2-fluorobenzothiazole, 2-fluorobenzo d thiazole, 2-benzothiazolyl fluoride, benzothiazole, 2-fluoro, 2-fluoro-benzothiazole, fluorobenzothiazole, acmc-2099en, #
PubChem CID 70753
IUPAC Name 2-fluoro-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)F

Spezifikation

Density 1.33
Boiling Point 97°C to 98°C (16 mmHg)
Odor Characteristic
Refractive Index 1.583
Quantity 1 g
UN Number UN2810
Beilstein 971503
Formula Weight 153.18
Percent Purity 99%
Chemical Name or Material 2-Fluorobenzothiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'