missing translation for 'onlineSavingsMsg'
Learn More
2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals Product Code.: 11414913

2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals

Product Code. 11414913
25 g
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 11414913

Brand: Thermo Scientific Alfa Aesar A12227.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.

Solubility
Soluble in alcohol, ether. Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-60-8
Molecular Formula C12H9F
Molecular Weight (g/mol) 172.20
MDL Number MFCD00000317
InChI Key KLECYOQFQXJYBC-UHFFFAOYSA-N
Synonym 2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene
PubChem CID 67579
SMILES FC1=CC=CC=C1C1=CC=CC=C1

Specifications

Melting Point 72°C to 76°C
Density 1.245
Boiling Point 248°C
Quantity 25 g
Beilstein 2043175
Solubility Information Soluble in alcohol,ether. Insoluble in water.
IUPAC Name 1-fluoro-2-phenylbenzene
Formula Weight 172.2
Percent Purity 98%
Chemical Name or Material 2-Fluorobiphenyl

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250790 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12227.14,A12227.06,A12227.09
Lot Number10245702 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA12227.14,A12227.06,A12227.09
Lot NumberX16G016 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberA12227.14,A12227.06,A12227.09
Lot NumberX28I003 Certificate TypeCertificate of Analysis Date11/11/2022 Catalog NumberA12227.14,A12227.06,A12227.09
Lot Number10153702 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12227.14,A12227.06,A12227.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.