missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorostyrene, 98%, stab. with 0.1% 4-tert-butylcatechol
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents, heat, light, UV radiation and free radical initiators. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
It finds its application in the homo- and cross-olefin metathesis coupling of vinylphosphane oxides and electron-poor alkenes.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents, heat, light, UV radiation and free radical initiators. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.
Specifications
Specifications
| Density | 1.02 |
| Boiling Point | 29°C to 30°C (4 mmHg) |
| Flash Point | 35°C (95°F) |
| Odor | Recognizable |
| Refractive Index | 1.52 |
| Quantity | 25 g |
| UN Number | UN1993 |
| Beilstein | 1925226 |
| Solubility Information | Not miscible or difficult to mix in water. |
| Formula Weight | 122.14 |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction