missing translation for 'onlineSavingsMsg'
Learn More

2-Furaldehyde, 99%

Product Code. 10686382 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
25mL
This item is not returnable. View return policy

Product Code. 10686382

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-01-1 | C5H4O2 | 96.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-01-1
Molecular Formula C5H4O2
Molecular Weight (g/mol) 96.09
MDL Number MFCD00003229
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
PubChem CID 7362
ChEBI CHEBI:34768
IUPAC Name furan-2-carbaldehyde
SMILES O=CC1=CC=CO1

Specifications

Acidity 0.02 eq/L max.
CAS Min % 98.5
CAS Max % 100.0
Melting Point -37°C
Color Amber to Brown
Density 1.1600g/mL
Boiling Point 159°C to 161°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5250 to 1.527
Quantity 250 mL
Residue after Evaporation 0.5% max.
Merck Index 15,4333
Specific Gravity 1.16
Solubility Information Solubility in water: 83g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
Viscosity 1.49 mPa.s (25°C)
Formula Weight 96.08
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Furaldehyde
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Furaldehyde
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.