missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid, 98%, Thermo Scientific Chemicals

Product Code. 10754231
500 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-14-2
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoovaShow MoreShow Less
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 129.0°C to 133.0°C
Color Beige to White
Boiling Point 230.0°C to 232.0°C
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Molecular Formula C5H4O3
Quantity 500 g
Beilstein 18, 272
Merck Index 15, 4336
Solubility Information Solubility in water: 36g/L (20°C). Other solubilities: soluble in alcohol and ether,insoluble in paraffin hydrocarbons
Molecular Weight (g/mol) 112.08
Formula Weight 112.08
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Furoic acid, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435038 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number119825000,119820050,119820025,119821000
Lot NumberA0406864 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number119825000,119820050,119820025,119821000
Lot NumberA0405355 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number119825000,119820050,119820025,119821000
Lot NumberA0452313 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number119825000,119820050,119820025,119821000
Lot NumberA0402266 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number119825000,119820050,119820025,119821000
compliance-icons compliance-icons
Product Identifier
  • 2-Furoic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Furoic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.