missing translation for 'onlineSavingsMsg'
Learn More

2-Heptylamine, 98+%, Thermo Scientific Chemicals

50.25 € - 538.68 €

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10764591
View Documents
Thermo Scientific Acros
103770250
25 mL Glass bottle
50.25 €
25mL
Please to purchase this item. Need a web account? Register with us today!
10112850
View Documents
Thermo Scientific Acros
103771000
100 mL Glass bottle
141.71 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10647953
View Documents
Thermo Scientific Acros
103775000
500 mL Glass bottle
538.68 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-82-0
115.22
VSRBKQFNFZQRBM-UHFFFAOYNA-N
5603
CCCCCC(C)N
C7H17N
MFCD00008101
2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
heptan-2-amine

Specifications

123-82-0
100.0
0.7680g/mL
54°C
98% min. (GC)
CH3(CH2)4CH(NH2)CH3
MFCD00008101
15,9982
2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
VSRBKQFNFZQRBM-UHFFFAOYNA-N
heptan-2-amine
5603
98+%
2-Heptylamine, ≥98%
98.5
Colorless to Yellow
142.0°C to 144.0°C
Authentic
C7H17N
1.4175 to 1.4195
04,194
0.768
Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol, ether, petroleum ether,, chloroform and benzene
CCCCCC(C)N
115.22
115.22
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rin

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-655-5

missing translation for 'rtecsNumber' : MQ5425000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Heptylamine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only