missing translation for 'onlineSavingsMsg'
Learn More

2-Heptyn-1-ol, 97%, Thermo Scientific Chemicals

Product Code. 11497157
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-36-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00039541
InChI Key OGDYROFIIXDTQK-UHFFFAOYSA-N
Synonym 2-heptyn-1-ol, 2-heptyne-1-ol, 2-heptynol, pubchem13065, acmc-2097no, hoch2c=c ch2 3ch3
PubChem CID 535111
IUPAC Name hept-2-yn-1-ol
SMILES CCCCC#CCO

Specifications

Density 0.882
Boiling Point 89°C to 90°C (16 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.454
Quantity 5 g
Beilstein 1739001
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material 2-Heptyn-1-ol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'