Learn More
2-Hexanone, 98%, Thermo Scientific Chemicals
Description
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (±)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Hexanone acts as a solvent for lacquers, ink thinners, nitrocellulose, resins and waxes. It is also employed in the preparation of (+/-)-monomorine. It also plays a useful role in the preparation of alcohols by transfer hydrogenation of ketones. Further, it is used as a medium evaporating solvent for nitrocellulose acrylates, vinyl and alkyd coatings.
Solubility
Miscible with acetone, ethanol and ether. Slightly miscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases and reducing agents.

Specifications
Specifications
Melting Point | -56°C |
Density | 0.811 |
Boiling Point | 126°C to 127°C |
Flash Point | 23°C (73°F) |
Odor | Pungent |
Refractive Index | 1.401 |
Quantity | 100 g |
UN Number | UN1224 |
Beilstein | 1737676 |
Merck Index | 14,6033 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462053 | Certificate of Analysis | 30/08/2024 | A14867.36,A14867.22,A14867.09,A1486701,A14867.18 |
A0442805 | Certificate of Analysis | 25/08/2023 | A14867.36,A14867.22,A14867.09,A14867.18 |
10169263 | Certificate of Analysis | 22/11/2021 | A14867.36,A14867.22,A14867.09,A14867.18 |
10146444 | Certificate of Analysis | 22/11/2021 | A14867.36,A14867.22,A14867.09,A14867.18 |
10181162 | Certificate of Analysis | 22/11/2021 | A14867.36,A14867.22,A14867.09,A14867.18 |
Safety and Handling



- 2-Hexanone
- Danger
- Flammable liquid Category 3
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H372-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 2-Hexanone
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.