Learn More
2'-Hydroxy-4',5'-dimethylacetophenone, 98%, Thermo Scientific Chemicals
41.20 € - 154.00 €
Chemical Identifiers
CAS | 36436-65-4 |
---|---|
Molecular Formula | C10H12O2 |
Molecular Weight (g/mol) | 164.204 |
MDL Number | MFCD00002306 |
InChI Key | YXVSURZEXVMUAM-UHFFFAOYSA-N |
PubChem CID | 118976 |
IUPAC Name | 1-(2-hydroxy-4,5-dimethylphenyl)ethanone |
SMILES | CC1=C(C=C(C(=C1)C(=O)C)O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11410240
|
Thermo Scientific Alfa Aesar
H50685.03 |
1 g |
41.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11420240
|
Thermo Scientific Alfa Aesar
H50685.06 |
5 g |
154.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2'-Hydroxy-4',5'-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Hydroxy-4′,5′-dimethylacetophenone is used in the preparation of chalcone derivatives with antibacterial activity.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
Chemical Identifiers
36436-65-4 | |
164.204 | |
YXVSURZEXVMUAM-UHFFFAOYSA-N | |
1-(2-hydroxy-4,5-dimethylphenyl)ethanone |
C10H12O2 | |
MFCD00002306 | |
118976 | |
CC1=C(C=C(C(=C1)C(=O)C)O)C |
Specifications
36436-65-4 | |
C10H12O2 | |
2'-hydroxy-4',5'-dimethylacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethanone, 2-hydroxy-4,5-dimethylacetophenone, 4'5'-dimethyl-2'-hydroxyacetophenone, ethanone, 1-2-hydroxy-4,5-dimethylphenyl, 4',5'-dimethyl-2'-hydroxyacetophenone, 1-2-hydroxy-4,5-dimethylphenyl ethan-1-one, 1-acetyl-2-hydroxy-4,5-dimethylbenzene, ethanone,1-2-hydroxy-4,5-dimethylphenyl, acmc-1ctvf | |
YXVSURZEXVMUAM-UHFFFAOYSA-N | |
1-(2-hydroxy-4,5-dimethylphenyl)ethanone | |
118976 | |
98% |
88°C to 90°C | |
MFCD00002306 | |
Slightly soluble in water. | |
CC1=C(C=C(C(=C1)C(=O)C)O)C | |
164.204 | |
164.21 | |
2'-Hydroxy-4',5'-dimethylacetophenone |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 253-035-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only