missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid hydrate, tech. 85%, may cont. up to 10% 2-propanol, Thermo Scientific Chemicals

Product Code. 11333436
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4065-45-6
Molecular Formula C14H12O6S
Molecular Weight (g/mol) 308.30
MDL Number MFCD00024962
InChI Key CXVGEDCSTKKODG-UHFFFAOYSA-N
Synonym sulisobenzone, benzophenone-4, sungard, 2-hydroxy-4-methoxybenzophenone-5-sulfonic acid, benzophenone 4, uval, uvinul, benzenesulfonic acid, 5-benzoyl-4-hydroxy-2-methoxy, sulisobenzona, sulisobenzonum
PubChem CID 19988
IUPAC Name 5-benzoyl-4-hydroxy-2-methoxybenzenesulfonic acid
SMILES COC1=CC(O)=C(C=C1S(O)(=O)=O)C(=O)C1=CC=CC=C1

Specifications

Melting Point 108°C to 112°C
Quantity 10 g
Beilstein 2889165
Merck Index 14,8983
Formula Weight 308.31 (Anhydrous)
Percent Purity 85%
Grade Technical
Assay may cont. up to 10% 2-propanol
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone-5-sulfonic acid hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.