missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 10011614
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93249-62-8
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00075249
InChI Key WQUZBERVMUEJTD-UHFFFAOYSA-N
PubChem CID 1268058
IUPAC Name 2-hydroxy-5-(trifluoromethoxy)benzaldehyde
SMILES C1=CC(=C(C=C1OC(F)(F)F)C=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 31.0°C to 33.0°C
Color White to Yellow
Boiling Point 82.0°C (60.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CF3OC6H3(OH)CHO
Quantity 250 mg
Synonym 2-hydroxy-5-trifluoromethoxy benzaldehyde, 5-trifluoromethoxy salicylaldehyde, benzaldehyde, 2-hydroxy-5-trifluoromethoxy, acmc-20antc, 5-trifluoromethoxysalicylaldehyde, 5-trifluoromethoxy-salicylaldehyde, 2-hydroxy-5-trifluoromethoxy-benzaldehyde, 20hydroxy-5-trifluoromethoxy benzaldehyde, 2-oxidanyl-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-hydroxy-5-trifluoromethoxy
Formula Weight 206.12
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'