missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Hydroxy-5-(trifluoromethoxy)benzonitrile, 95%

Product Code. 11382069
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 875664-40-7
Molecular Formula C8H4F3NO2
Molecular Weight (g/mol) 203.12
MDL Number MFCD04973781
InChI Key GOAVRYAOBXORSY-UHFFFAOYSA-N
Synonym 2-hydroxy-5-trifluoromethoxy benzonitrile
PubChem CID 24721643
IUPAC Name 2-hydroxy-5-(trifluoromethoxy)benzonitrile
SMILES C1=CC(=C(C=C1OC(F)(F)F)C#N)O

Specifications

Melting Point 84°C to 90°C
Quantity 1 g
UN Number UN3439
Formula Weight 203.12
Percent Purity 95%
Chemical Name or Material 2-Hydroxy-5-(trifluoromethoxy)benzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'