missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-6'-methoxyacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 11458457
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 703-23-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00008732
InChI Key UENLHUMCIOWYQN-UHFFFAOYSA-N
PubChem CID 69709
IUPAC Name 1-(2-hydroxy-6-methoxyphenyl)ethanone
SMILES COC1=CC=CC(O)=C1C(C)=O

Specifications

Melting Point 56°C to 59°C
Boiling Point 141°C (16 mmHg)
Quantity 5 g
Beilstein 1869049
Synonym 1-2-hydroxy-6-methoxyphenyl ethanone, 2'-hydroxy-6'-methoxyacetophenone, 2-hydroxy-6-methoxyacetophenone, 1-2-hydroxy-6-methoxyphenyl ethan-1-one, ethanone, 1-2-hydroxy-6-methoxyphenyl, acetophenone, 2'-hydroxy-6'-methoxy, unii-m5vqg8968n, ethanone,1-2-hydroxy-6-methoxyphenyl, 1-2-hydroxy-6-methoxy-phenyl-ethanone, 1-acetyl-2-hydroxy-6-methoxybenzene
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2'-Hydroxy-6'-methoxyacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'