missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxychalcone, 95%, Thermo Scientific Chemicals

Product Code. 11348835
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1214-47-7
Molecular Formula C15H12O2
Molecular Weight (g/mol) 224.259
MDL Number MFCD00016441
InChI Key AETKQQBRKSELEL-ZHACJKMWSA-N
Synonym 2'-hydroxychalcone, o-hydroxychalcone, e-2'-hydroxychalcone, 1-2-hydroxyphenyl-3-phenylprop-2-en-1-one, chalcone, 2'-hydroxy, ccris 7796, 1-2-hydroxyphenyl-3-phenyl-2-propen-1-one, unii-vy06dz94oc, acrylophenone, 2'-hydroxy-3-phenyl, 2-propen-1-one, 1-2-hydroxyphenyl-3-phenyl
PubChem CID 638276
ChEBI CHEBI:27916
IUPAC Name (E)-1-(2-hydroxyphenyl)-3-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C=CC(=O)C2=CC=CC=C2O

Specifications

Melting Point 89°C to 91°C
Quantity 5 g
Beilstein 976324
Formula Weight 224.26
Percent Purity ≥98%
Chemical Name or Material 2'-Hydroxychalcone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'