missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl acrylate, 97%, stabilized

Product Code. 10309090 Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
5mL
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10309090

Brand: Thermo Scientific undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 818-61-1 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-61-1, 150-76-5
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002865
InChI Key OMIGHNLMNHATMP-UHFFFAOYSA-N
Synonym 2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
PubChem CID 13165
IUPAC Name 2-hydroxyethyl prop-2-enoate
SMILES C=CC(=O)OCCO

Specifications

Acidity 4mg KOH/g max.
CAS Min % 96.0
CAS Max % 100.0
Melting Point -60°C
Color Yellow to Brown
Density 1.1060g/mL
Boiling Point 210°C to 215°C
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.451
Quantity 2.5 L
Specific Gravity 1.106
Solubility Information Solubility in water: soluble in water
Stabilizer 200 to 450ppm MEHQ
Viscosity 9 mPa.s (20°C)
Formula Weight 116.12
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material 2-Hydroxyethyl acrylate, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Hydroxyethyl acrylate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.